ContactUsLinkComponent
Detergent
NEUTRAD® Neutral pH Liquid Cleaner, Decon Labs
Catalog #: CA89234-804
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Detergent
NEUTRAD® Neutral pH Liquid Cleaner, Decon Labs
Catalog #: CA89234-804
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Size:
    5 L (1.32 gal.)
  • Cat. no.:
    CA89234-804
  • Packaging:
    Poly Bottle
  • Supplier No.:
    3001
Specifications
About this item
NEUTRAD® is a safe detergent for manual scrub cleaning of any labware – replaces old fashioned caustic soap powders! Highly concentrated, very economical. Near-neutral, pH is skin-safe! Phosphate-free, very rinsable and biodegradable. Liquid NEUTRAD® is instantly soluble, totally rinsable, safe, near-neutral pH and phosphate-free. Made with recycled packaging.
Customers Who Bought This Also Bought