ContactUsLinkComponent
Detergent
Contrad NF, Non-foaming Liquid Detergent, Decon Labs
Catalog #: CA89233-164
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Detergent
Contrad NF, Non-foaming Liquid Detergent, Decon Labs
Catalog #: CA89233-164
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Size:
    3.8 L (1.0 gal.)
  • Cat. no.:
    CA89233-164
  • Packaging:
    Poly Bottle
  • Supplier No.:
    6002
Specifications
About this item
Contrad NF is a non-foaming concentrated liquid detergent designed for high-pressure cleaning applications – laboratory and industrial automatic washing machines, CIP systems, steam washers, etc
Phosphate-free and chlorine-free, Contrad NF is particularly recommended for critical cleaning of labware, where total rinsability and residue-free results are vital; and for plant cleaning, such as biotechnology and pharmaceutical production equipment. Made with recycled packaging.
Customers Who Bought This Also Bought