Order Entry
ContactUsLinkComponent
Detergent
Contrad 70, liquid detergent, Decon Labs
Catalog #: CA89233-152
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Detergent
Contrad 70, liquid detergent, Decon Labs
Catalog #: CA89233-152
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Size:
    1 L (34 oz.)
  • Cat. no.:
    CA89233-152
  • Packaging:
    Poly Bottle
  • Supplier No.:
    1002

Specifications

About this item

Contrad 70 liquid detergent concentrate is phosphate-free, chlorine-free, and biodegradable

Cleans glass, plastic, ceramic, and ferrous metals by soaking, manual scrubbing, or ultrasonic cleaning. Contrad 70 is free-rinsing and disposable straight to the drain. Made with recycled packaging.

Customers Who Bought This Also Bought