Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Catalog #: CA1.05432.1011
Supplier: MilliporeSigma
CAS Number: 1336-21-6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Size:1 L
- Cat. no.:CA1.05432.1011
- Supplier No.:1.05432.1011
Specifications
Specification Test Results
- Assay (acidimetric, NH₃):25.0 - 30.0 %
- Identity:passes test
- Appearance of solution:passes test
- Density (20/20°C):0.892 - 0.910
- Carbonate (as CO₂):≤ 10 ppm
- Chloride (Cl):≤ 0.5 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Silicon (as SiO₂):≤ 10 ppm
- Sulfate (SO₄):≤ 2 ppm
- Sulphide (S):≤ 0.2 ppm
- Pyridine and related substances:≤ 2 ppm
- Ag (Silver):≤ 0.020 ppm
- Al (Aluminium):≤ 0.500 ppm
- Au (Gold):≤ 0.100 ppm
- Ba (Barium):≤ 0.050 ppm
- Bi (Bismuth):≤ 0.100 ppm
- Ca (Calcium):≤ 0.500 ppm
- Cd (Cadmium):≤ 0.050 ppm
- Co (Cobalt):≤ 0.050 ppm
- Cr (Chromium):≤ 0.050 ppm
- Cu (Copper):≤ 0.100 ppm
- Fe (Iron):≤ 0.100 ppm
- Ga (Gallium):≤ 0.020 ppm
- In (Indium):≤ 0.020 ppm
- K (Potassium):≤ 0.500 ppm
- Li (Lithium):≤ 0.020 ppm
- Mg (Magnesium):≤ 0.100 ppm
- Mn (Manganese):≤ 0.050 ppm
- Mo (Molybdenum):≤ 0.050 ppm
- Na (Sodium):≤ 0.500 ppm
- Ni (Nickel):≤ 0.050 ppm
- Pb (Lead):≤ 0.050 ppm
- Pt (Platinum):≤ 0.100 ppm
- Sn (Tin):≤ 0.100 ppm
- Sr (Strontium):≤ 0.100 ppm
- Ti (Titanium):≤ 0.100 ppm
- Tl (Thallium):≤ 0.050 ppm
- Zn (Zinc):≤ 0.100 ppm
- Oxidizable substances:passes test
- Substances reducing potassium permanganate (as O):≤ 5 ppm
- Residue on ignition (as SO₄):≤ 10 ppm
- Non volatile matter:≤ 10 ppm
- Supelco®, Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis:
Specification Test Results
About this item
Synonyms: Ammonia aqueous,Ammonium hydroxide solution,Ammonium hydroxide
:Ammonia solution
Boiling Pt:37.7 °C (1013 hPa)
Melting Pt:–57.5 °C
Density:0.9 g/ml (25°C) (lit.)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 37.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –57.5 °C
:Flash Pt:
:Flash Pt:
:M.W. 35.05 g/mol
EINECS:215-647-6
:37.7 °C (1013 hPa)
Storage Temperature:Ambient
UN:2672
:8
:III
:GHS05
:GHS07
:GHS09
Customers Who Bought This Also Bought