Order Entry
ContactUsLinkComponent
Reagent Reservoir with Divider, Polystyrene, 25mL, Sterile, 0.84 oz
Matrix™ Reagent Reservoirs, Thermo Scientific
Catalog #: CA101505-066
Matrix™ Reagent Reservoirs, Thermo Scientific
Catalog #: CA101505-066

Specifications

  • Capacity:
    25 ml
  • Sterility:
    Sterile
  • Color:
    White
  • Description:
    Reagent Reservoir with Divider
  • Material:
    Polystyrene
  • Cat. no.:
    CA101505-066
  • Packaging:
    10 Bags of 10
  • Supplier No.:
    8095

Specifications

About this item

Matrix™ Reagent Reservoirs are sterile and offer an extra-wide base to add rigidity and stability, helping to avoid spills.

  • Trough within a trough maximizes amount of liquid accessible to pipet tips
  • Pour-off spouts on all four corners reduce spillage
  • Graduations on inside wall enable quick measurement
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Customers Who Bought This Also Bought