ContactUsLinkComponent
Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040in Red PTFE/Silicone Septa Lined BONDED CLOSURE With Slit. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog #: 10171-060
Supplier:  Chemglass
Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040in Red PTFE/Silicone Septa Lined BONDED CLOSURE With Slit. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040in Red PTFE/Silicone Septa Lined BONDED CLOSURE With Slit. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog #: 10171-060
Supplier:  Chemglass
Customers Who Bought This Also Bought