Vial, 2.0mL, Amber with Marking Spot, Silanized, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Catalog #: 10171-014
Supplier: Chemglass
Customers Who Bought This Also Bought