Fixed height sample tube for 7l bioreactors, M10 thread, max immersion length: 315mm
Catalog #: 10170-830
Supplier: Chemglass
Fixed height sample tube for 7l bioreactors, M10 thread, max immersion length: 315mm
Catalog #: 10170-830
Supplier: Chemglass
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
1 747,88 $
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
362,33 $
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
240,47 $ to 8 653,27 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
359,30 $
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
121,35 $ to 993,13 $
Chemware® Laboratory Matting, PTFE, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
1 947,60 $