Media Bottles, Anerobic, Clear, Chemglass
Catalog #: 10170-766
Supplier: Chemglass
Small Business Enterprise
Specifications
- Cap Type:Aluminium Seal
- Capacity:1000 ml
- Description:Anaerobic Bottles
- Cat. no.:10170-766
- Supplier No.:CLS-4217-05
Specifications
About this item
Anaerobic media bottles are designed for maintaining anaerobic conditions, useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development.
- Autoclavable
- Manufactured from Duran® Schott borosilicate glass
Each bottle has a graduation scale indicating the approximate volumes and has a writing area for identification. Bottle is supplied with a rubber stopper and aluminum seal.
Customers Who Bought This Also Bought