ContactUsLinkComponent
Kit, Vial, 2.0mL, Clear with Spot, 12 x32mm, with 0.30mL Fused Insert.040in PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog #: 10170-586
Supplier:  Chemglass
Kit, Vial, 2.0mL, Clear with Spot, 12 x32mm, with 0.30mL Fused Insert.040in PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Kit, Vial, 2.0mL, Clear with Spot, 12 x32mm, with 0.30mL Fused Insert.040in PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog #: 10170-586
Supplier:  Chemglass
Customers Who Bought This Also Bought