Septum Stoppers, Sleeve Type, Chemglass
Catalog #: 10170-488
Supplier: Chemglass
Small Business Enterprise
Specifications
- Color:Red
- Description:Stopper, Sleeve Type, for 19/22 to 19/38 Outer Joints
- Cat. no.:10170-488
- Supplier No.:CG-3022-96
Specifications
About this item
Sleeve type stoppers with hollow plugs and sleeve extensions.
- Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Customers Who Bought This Also Bought