Order Entry
ContactUsLinkComponent
Bottle, Media, Glass, 5l, Duran, Clear, With hose barb and GL-45 vented cap
DURAN® Media Storage Bottles with Hose Barb, Clear Glass, Chemglass
Catalog #: 10170-338
Supplier:  Chemglass
Small Business Enterprise
Bottle, Media, Glass, 5l, Duran, Clear, With hose barb and GL-45 vented cap
DURAN® Media Storage Bottles with Hose Barb, Clear Glass, Chemglass
Catalog #: 10170-338
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Cap Size:
    GL 45
  • Capacity:
    5000 ml (169 oz.)
  • Description:
    Bottle, with hose barb for 1/4" tubing
  • Cat. no.:
    10170-338
  • Supplier No.:
    CLS-1423-5L

Specifications

About this item

Use these clear Duran® media bottles for easy gravity transfer of solutions.

  • Drip-free pouring ring
  • One-piece, liner-less vented cap is autoclavable and reusable

The hose barb outlet at the bottom allows for easy gravity transfer of solutions.

Customers Who Bought This Also Bought