Order Entry
ContactUsLinkComponent
Adjustable sampling tube, 232mm maximum immersion length, For 3l and 5l bio reactors
Adjustable Sampling Tubes for Bioreactors, M10
Catalog #: 10170-320
Supplier:  Chemglass
Small Business Enterprise
Adjustable sampling tube, 232mm maximum immersion length, For 3l and 5l bio reactors
Adjustable Sampling Tubes for Bioreactors, M10
Catalog #: 10170-320
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Length:
    232 mm
  • Volume:
    2 - 5 L
  • Description:
    Adjustable sampling tube
  • Supplier No.:
    CLS-1380-21

Specifications

About this item

These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.

Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.

Customers Who Bought This Also Bought