Crimpers, DWK Life Sciences
Catalog #: 10158-524
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Aluminum seal manual crimper
- Seal Size:11 mm
- Supplier No.:69902-11
Specifications
About this item
Manual hand crimpers for aluminum seals and vials.
Perfect for chromatography and general crimping needs.
Customers Who Bought This Also Bought