To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Forkhead box D4
- Antigen symbol:FOXD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen synonyms:Forkhead box protein D4-like 2|FOXD4-like 3|Forkhead box protein D4-like 4|FOXD4-like 5|Forkhead box protein D4-like 5|FOXD4-like 4|Forkhead box protein D4-like 6|Forkhead box protein D4-like 3|FOXD4-like 6|FOXD4-like 2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:FOXD4
- Purification:Antigen affinity purification
- Cat. no.:10156-102
- Supplier No.:24835-1-AP
The FOXD4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FOXD4. This antibody recognizes human, mouse, rat antigen. The FOXD4 antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 600 ; IHC 1 : 50
Type: Primary
Antigen: FOXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat