To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:500 ml (16.9 oz.)
- Sterility:Non sterile
- Material:PP
- Supplier No.:6542
Specifications
About this item
These large capacity containers are available in 8 oz., 16 oz., and 32 oz. sizes.
Containers are bulk packaged with separate yellow caps.
The screwcaps provide a tight seal and are ribbed for easy opening and closing. The containers are perfect for long-term storing of liquids, solids, and large histology specimens.