Autoclave Indicator Tape, Lead Free, Micronova
Catalog #: 10153-154
Supplier: Micronova
Small Business Enterprise
Specifications
- Dimensions:2" x 60 yds
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:10153-154
- Supplier No.:AUTOPC-2
Specifications
About this item
Method of positively confirming complete sterilization cycle in both gravity discharge and vacuum autoclaves.
- Lead and latex-free
- Wound on plastic core
- Easy to handle tape
The green indicator lines change to black when proper levels of steam pressure and temperature have been achieved. The uniquely printed lines cannot be rubbed off and will not contaminate instruments. After use, the tape removes quickly and completely, leaving minimal residue or staining. Tape adheres on contact and stays in place through live steam pressure.
Certifications: RoHS compliant.
Customers Who Bought This Also Bought