Vial with snap cap clear 40DR case 100
Catalog #: CA10152-870
Supplier: Richards Packaging
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$150.16 to $1,502.77
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
Supplier: MilliporeSigma
$172.55
Sodium chloride ≥99.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$110.33 to $110.33