Pregnancy Tests, Stanbio Laboratory
Catalog #: 10152-682
Supplier: Stanbio Laboratory
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:QuPID® hCG (Urine)
- Size:25 Test
- Cat. no.:10152-682
- Sensitivity:20 mIU/mL
- Certification:FDA, CE, Health Canada
- Supplier No.:1220-025
- No. of Tests:25
Specifications
About this item
Stanbio Laboratory offers pregnancy tests to meet your clinical and laboratory needs.
Sensivity in Urine 20 mIU/mL
Sensivity in Serum 10 mIU/mL (20 mIU/mL for the True® 20 Plus set)
Certifications: All pregnancy tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: Tests are single step cassette platform formats for urine only or urine/serum pregnancy testing.
Customers Who Bought This Also Bought