Accessories for Vileda Professional® Controlled Environment Trolleys
Catalog #: 10153-174
Supplier: Vileda Professional - FHP
Specifications
- For Use With:CE Bucket Trolley or CE Pre-Prepared Trolley; Can be used for the pre-prepared method or as storage
- Description:Pre-Prepared Box, 15 x 11 x 8", Blue
- Cat. no.:10153-174
- Supplier No.:120774
- Details:Easily stacked; Color coding clips included to allow differentiation in colors of buckets (red, blue, yellow, green); Lids (sold separately) snaps on to the Pre-Prepared Box easily and securely; Sieve (sold separately) included evenly placed holes which ensure even saturation of mops; Autoclavable