Order Entry
ContactUsLinkComponent
BUFFER CHARC YEAST EXTRACT PLATE PK10
BCYE Selective Agar with GPCV Antibiotics, Hardy Diagnostics
Catalog #: CA10142-512
CAS Number:  
BUFFER CHARC YEAST EXTRACT PLATE PK10
BCYE Selective Agar with GPCV Antibiotics, Hardy Diagnostics
Catalog #: CA10142-512
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Plate
  • Description:
    BCYE w/GPCV
  • Size:
    15 x 100 mm
  • Volume:
    26 ml
  • Supplier No.:
    W169

Specifications

About this item

For the selective isolation of Legionella spp.

  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Optically clear polystyrene for distortion free viewing

Buffered Charcoal Yeast Extract Agar with Glycine, Vancomycin, Polymyxin, and Cycloheximide) for the selective isolation of Legionella spp.

Conforms to ISO11371 formulation.

Customers Who Bought This Also Bought