Order Entry
Canada
ContactUsLinkComponent
ß-Hydroxybutyrate LiquiColor® Test (Endpoint), LiquiColor® & Liqui-UV®
ß-Hydroxybutyrate LiquiColor® Test, Stanbio Laboratory
Catalog # 10152-474
Supplier:  Stanbio Laboratory
CAS Number:  
ß-Hydroxybutyrate LiquiColor® Test, Stanbio Laboratory
Catalog # 10152-474
Supplier:  Stanbio Laboratory
Supplier Number:  2440-058
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ß-Hydroxybutyrate LiquiColor® Test (Endpoint)
  • Environmentally Preferable:
  • Cat. no.:
    10152-474
  • Contents:
    Reagent 1: 1 x 50 mL; Reagent 2: 1 x 8.5 mL; Standard (1 mmol/L): 1 x 3 mL
  • Supplier No.:
    2440-058

Specifications

About this item

Quantitative determination of ß-Hydroxybutyrate in human serum or plasma.

  • Available in two kit configurations
  • Determine presence and degree of ketosis
  • Linearity 4.5 mmol/L (46.8 mg/dL)

For the Quantitative determination of ß-Hydroxybutyrate in human serum or plasma. The presence and degree of ketosis can be determined by measuring blood levels of ß-hydroxybutyrate. This reagent is available in two kit configurations: a dual reagent kit for open channel chemistry analyzers and a 2 x 90 test kit that may be used directly on the Beckman Synchron® CX, LX and DX analyzers. Both kits include a 3 mL standard. Instrument specific applications are available from the manufacturer for open-channel automated and semi-automated chemistry analyzers. The ß-Hydroxybutyrate Reagent can also be used in a five minute procedure utilizing a spectrophotometer which reads absorbance at 505 nm wavelength.

Certifications: FDA 510(k). CE marked.
Ordering information: ß-Hydroxybutyrate tri-level control set, bi-level control set, and a linearity standard set are sold separately. A ß-Hydroxybutyrate validation pack and a Method Comparison kit are offered as resources to help laboratories get started using the ß-Hydroxybutyrate LiquiColor® test.
Caution: Store at 2 - 8º C until the expiration date on respective labeling.
Frequently Bought Together