Order Entry
ContactUsLinkComponent
FILTER FUNNEL SENTINO 100ML CS100
Sentino® Filter Funnels, Cytiva (Formerly Pall Lab)
Catalog #: CA10147-760
Supplier:  Cytiva
FILTER FUNNEL SENTINO 100ML CS100
Sentino® Filter Funnels, Cytiva (Formerly Pall Lab)
Catalog #: CA10147-760
Supplier:  Cytiva

Specifications

  • Dimensions:
    8.1D x 5.3H cm (3.2 x 2.1")
  • Volume:
    100 ml
  • Description:
    Sentino® Filter Funnels
  • Material:
    PP
  • Supplier No.:
    4870

Specifications

About this item

The Sentino® filter funnel and adapter are designed with a press-fit seal, membrane guides and forceps access point allowing for simple set-up and membrane removal. This will speed up the process time in the laboratory.

  • Easy placement
  • Risk reduction
  • Regulatory compliance
  • Allows easy and accurate sample measurement
  • Available in 100 and 250 ml capacity to accommodate a variety of sample sizes
  • Inert polypropylene construction to preserve the integrity of the sample
  • Base adapts for use with Sentino® microbiology pump or standard manifold

Easy to use, disposable funnels for microbiological monitoring of aqueous solutions.

The Sentino® filter funnel and Pall membranes reduces the risk of a contamination event as compared to reusable funnels. The filter funnels are pre-sterilised via gamma sterilisation eliminating the need for autoclaving or any other cleaning/sanitising techniques. This will save time and money by eliminating the need to investigate contamination events resulting from the reusable devices. Membrane filtration is the method of choice for microbial monitoring of aqueous solutions as specified in 'Standard Methods for the Examination of Water and Wastewater' and Pharmacopoeias worldwide. The funnels are marked in three increments (20 ml, 50 ml and 100 ml) and are marked in four increments (50 ml, 100 ml, 200 ml and 250 ml) for higher visibility.

Ordering information: Accessories are required and sold separately for use with Sentino® filter funnels. They are supplied non sterile.
Customers Who Bought This Also Bought