Incubation Trays, Azure Biosystems
Catalog #: 10147-316
Supplier: Azure Biosystems
Product available on GSA Advantage®
Specifications
- Dimensions:7 x 5 cm
- Color:Black
- Description:Incubation Tray
- Cat. no.:10147-316
- Includes:5 Trays
- Supplier No.:AC2120
Specifications
About this item
Azure's specially designed incubation trays are ideal for staining and washing electrophoresis gels and membranes.
The trays have smooth interiors to avoid damage to gels or blots, and lids to protect them from dust.
The opaque trays are ideal for light-sensitive applications. Available in a range of sizes.
Customers Who Bought This Also Bought