To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Set of 3 Indicator Solutions
- Size:25 mL Dropper Bottle
- Chemical/Physical Specifications:Solutions
- Cat. no.:470301-296
- Grade:Laboratory
- Supplier No.:IX0003-SET/3
Specifications
About this item
One 25 mL bottle of each: Universal Indicator, Bromothymol Blue 0.04% (Aqueous), and Phenolphthalein 1% (Alcoholic).
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Flammable
Shelf Life: 1 year
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.