SODA ASH DENSE 12.5KG PLASTIC PAIL
Catalog #: CA10144-652
Supplier: Avantor
Customers Who Bought This Also Bought
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$99.85 to $385.36
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
$2,699.72
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$412.78 to $412.78