Order Entry
ContactUsLinkComponent
Gram Stain Set, contains 8oz each Crystal Violet, Stabilized Iodine, Decolorizer and Safranin and is used in the Gram staining of bacteria, 1 set.
VWR® Gram Stain Sets and Reagents
Catalog #: 10143-178
Supplier:  VWR International
CAS Number:  
Gram Stain Set, contains 8oz each Crystal Violet, Stabilized Iodine, Decolorizer and Safranin and is used in the Gram staining of bacteria, 1 set.
VWR® Gram Stain Sets and Reagents
Catalog #: 10143-178
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Gram Stain Set Stabilized Iodine
  • Size:
    4 x 237 mL (8 oz.)
  • Cat. no.:
    10143-178

Specifications

About this item

Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.

  • Sets include all necessary solutions
  • Enables differentiation

All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.

Customers Who Bought This Also Bought