To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Mineral Oil
- Size:500 mL
- Chemical/Physical Specifications:Heavy
- Cat. no.:470301-822
- Grade:Laboratory
- Supplier No.:MM0586-500ML
- Disposal Method:This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.
This material can only be disposed properly by a licensed hazardous waste disposal firm. Check to see if your school or district has an established relationship with a disposal firm and if not, check with a local college/university or other high schools in the area to see who they use to dispose of their hazardous waste.
Specifications
About this item
CAS Number: 8042-47-5
Formula Weight: Variable
Formula: CH2[CH2]nCH3
Density (g/mL): 0.818-0.880
Solubility: Organic Solvents
Synonyms: White Mineral Oil, Light Paraffin Oil
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.