Order Entry
ContactUsLinkComponent
Sodium Sulfide 9-Hydrate Reagent, 100 g
Sodium Sulfide 9-Hydrate
Catalog #: 470302-706
Supplier:  Ward's Science
CAS Number:  
Sodium Sulfide 9-Hydrate Reagent, 100 g
Sodium Sulfide 9-Hydrate
Catalog #: 470302-706
Supplier:  Ward's Science
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Sodium Sulfide 9-Hydrate
  • Size:
    100 g
  • Chemical/Physical Specifications:
    Crystal
  • Cat. no.:
    470302-706
  • Grade:
    ACS
  • Supplier No.:
    SS0846-100GR
  • Disposal Method:
    This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.

    Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.

     1. Prepare a 0.5 M sodium hydroxide solution and dissolve the sodium sulfide compound in the solution.
      a. Use about 100 mL NaOH for every 10 mL of sodium sulfide
     2. Slowly add sodium hypochlorite solution (bleach). Add 200 mL bleach for every 5 mL of sodium sulfide.
     3. After allowing the solution to sit overnight, flush the mixture down the drain with at least a 20-fold excess of water.
  • Solution Preparation:
    To prepare a solution, determine the formula weight of the material, the concentration (Molarity) of the solution, and the amount of solution desired in Liters. Once this information has been obtained, use the following formula:
    (Formula Weight) x (Concentration) x (Desired Volume) = Number of grams in desired volume of liquid

    For a 1 M, 1 L solution:
    (Formula Weight) x (1 mol/L) x (1 L) = x g added to 1-L volumetric flask. Dilute to a final volume of 1 L with DI Water.

Specifications

About this item

CAS Number: 1313-84-4
Formula Weight: 240.18
Formula: Na2S·9H2O
Density (g/mL): 1.43
Freezing Point (°C): 50
Solubility: Water and Alcohol
Synonyms: Sodium Monosulfide Nonahydrate
Shelf Life (months): 12
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Customers Who Bought This Also Bought