FectoPRO® DNA Transfection Kit for Protein Production, Transfection®
Catalog #: CA10118-842
Supplier: Polyplus Transfection
CAS Number:
Specifications
- Bioproduction:Yes
- Cell specific:HEK-293, CHO
- DNA:Yes
- siRNA:Yes
- Description:FectoPRO® DNA transfection kit
- Size:1 ml
- Cat. no.:CA10118-842
- Buffer Volume:1 ml
- Supplier No.:116-001
Specifications
About this item
The FectoPRO® Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various serum-free media, using low DNA amount (\<1 μg/ml of cell culture).
- Amazing protein and antibody production yields
- Cost-effective Transient Gene Expression using lower plasmid DNA amount
- Compatible with various mammalian expression media and cells systems
- Compliant with biomanufacturing guidelines
- Reproducible production yields
The FectoPRO®-mediated transfection process is easily scalable from a few milliliters to several liters of cell culture, ensuring robust reproducible protein production.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
FectoPRO® is guaranteed free of animal-origin components, and is chemically defined. Systematic lot management and release testing is performed for each lot produced. Furthermore FectoPRO® transfection kit undergoes advanced quality controls: protein productivity, cell viability, and complete sterility.
FectoPRO® is guaranteed free of animal-origin components, and is chemically defined. Systematic lot management and release testing is performed for each lot produced. Furthermore FectoPRO® transfection kit undergoes advanced quality controls: protein productivity, cell viability, and complete sterility.
Delivery: Each kit contains equal parts FectoPRO® Reagent and FectoPRO® Booster. FectoPRO® Booster is also available separately.
Customers Who Bought This Also Bought