To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:PBS (10X), pH 7.4
- Size:500 ml
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- Test description:Endotoxin and mycoplasma tested
- Shelf life:24 months
- Osmolality:300 ±5% mOsm/Kg H₂O
- Shipping temperature:Ambient
- Supplier No.:119-069-101
Specifications
About this item
PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
- Endotoxin and Performance tested
PBS is also useful in immunological applications, such as Western Blotting.