ContactUsLinkComponent
DPBS 1X
Dulbecco's Phosphate Buffered Saline (DPBS), Quality Biological
Catalog #: 10128-718
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
DPBS 1X
Dulbecco's Phosphate Buffered Saline (DPBS), Quality Biological
Catalog #: 10128-718
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    DPBS, 1X without Ca and Mg
  • Size:
    1 L
  • Sterility:
    Sterile filtered
  • Shipping temperature:
    Ambient
  • Supplier No.:
    114-057-131
Specifications
About this item
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
  • Without phenol red
DPBS can be used to provide a buffer system to maintain cell culture media in the physiological pH range of 7.0 to 7.6
DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time.
A 1X working solution is recommended for application usage.
DPBS is also used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate, to wash the wells of microtiter plates, and to wash cells during preparation and serial transfer.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought