Order Entry
ContactUsLinkComponent
Iscove's Modified Dulbecco's Medium
Iscove's Modified Dulbecco's Medium (IMDM), Quality Biological
Catalog #: 10128-638
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Iscove's Modified Dulbecco's Medium
Iscove's Modified Dulbecco's Medium (IMDM), Quality Biological
Catalog #: 10128-638
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    IMDM with L-glutamine and phenol red
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Shipping conditions:
    Ambient temperature
  • Osmolality:
    280 ±5% mOsm/Kg H₂0
  • pH:
    7.1 ±0.2
  • Shelf Life:
    24 months
  • Supplier No.:
    112-035-101

Specifications

About this item

Iscove’s Modified Dulbecco’s Medium was developed by Dr. Iscove and his coworkers to assist in their studies of hematopoietic stem cells. They modified Dulbecco’s MEM by the addition of HEPES buffer, selenium, additional amino acids, vitamins and sodium pyruvate, and potassium nitrate instead of ferric nitrate.

  • With L-glutamine and Phenol Red

IMDM is a highly enriched synthetic media well suited for rapidly proliferating, high-density cell cultures. Use IMDM to replace serum containing medium.

IMDM is a highly enriched synthetic media well suited for rapidly proliferating, high-density cell cultures. Use IMDM to replace serum containing medium.



Sterile Filtered
pH: 7.1 ± 0.2
Osmolality: 280±5%mOsm/Kg H20
Harmonization Code: 3821.00.0000
ECCN: EAR99

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Customers Who Bought This Also Bought