To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:McCoy’s 5A medium with L-Glutamine
- Size:500 ml
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- Osmolality:290 ±5% mOsm/Kg H₂O
- pH:7.0 ±0.2
- Shelf Life:24 months
- Supplier No.:112-028-101
Specifications
About this item
McCoy’s 5A Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. McCoy’s 5A Medium uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
- With L-glutamine and Phenol Red
McCoy’s 5A Medium originated from Dr. McCoy and his coworkers, as it was centered on the amino acid requirements of Novikoff hepatoma cells in culture. This work involved a medium known as Basal Medium 5A and used to grow normal bone marrow, skin, gingiva, testes, mouse kidney, omentum, adrenal glands, lung, spleen, rat embryos and other tissues. This product also includes Dr. Hsu’s addition of Hanks’ salts to enable use outside a CO2 incubator.
McCoy’s 5A Medium with L-glutamine will support growth of primary mammalian cells derived from the following: normal bone marrow skin, spleen, kidney, lung, rat embryos, and other tissues.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
A 10% FBS supplementation is recommended.