To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:EMEM with L-Glutamine
- Size:10× 500 ml
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- pH:7.0 ±0.2
- Shelf Life:24 months
- Supplier No.:112-018-101CS
Specifications
About this item
Eagle’s minimal essential medium (EMEM) is a cell culture medium developed by Harry Eagle that can be used to maintain cells in tissue culture.
- Basal medium with different variations.
MEM was developed by Dr. Harry Eagle to overcome problems encountered when attempting to grow normal fibroblasts and certain sub-types of HeLa cells. Many other modification of MEM followed including alpha- MEM, MEM Spinner and EMEM.
MEM Spinner was developed when calcium component was removed from the MEM medium, and resulted in a medium that supports the growth of cells in suspension.
Alpha MEM is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.
EMEM (Eagle’s Minimum Essential Medium) has a higher amino acid concentration and 2 to 5 times greater vitamin concentration than BME, permitting longer periods between feedings.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99