Order Entry
ContactUsLinkComponent
TNE Buffer
TNE Buffer, Quality Biological
Catalog #: 10128-582
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
TNE Buffer
TNE Buffer, Quality Biological
Catalog #: 10128-582
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • pH:
    7.8 - 8.0
  • Description:
    TNE buffer
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Test description:
    Endotoxin and mycoplasma tested
  • Shelf life:
    36 months
  • pH:
    7.8 - 8.0
  • DNase/RNase activity:
    Not detectable
  • Protease assay:
    Not detectable
  • Shipping temperature:
    Ambient
  • Supplier No.:
    351-302-101

Specifications

About this item

TNE Buffer is often used as a base material to generate lysis buffers.

  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested

More specifically, TNE Buffer is useful for: isolation of proteins from yeast cells as a lysis buffer, resuspension of pelleted virus or reporter virus particles and the flurometric determination of DNA content.

Supplier recommendation:

Without correcting the pH, concentrated virus can be used in non-denaturing SDS/PAGE-Western Blot analysis.
If the pH is corrected to inhibit triggering of the viral fusion machinery (for West Nile Virus, this corresponds to pH 7.4), the concentrated virus will remain infectious.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: This product requires special production. Please allow 2 to 4 weeks for delivery.
Caution: Research use only.
Customers Who Bought This Also Bought