TBS 10X Buffer Solution, pH 7.4, Quality Biological
Catalog #: 10128-546
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- pH:7.4 ±0.1
- Description:TBS 10X pH 7.4
- Size:500 ml
- Sterility:Sterile filtered
- Test description:DNAse, RNAse and protease tested
- Shelf life:36 months
- Shipping temperature:Ambient
- Supplier No.:351-086-101
Specifications
About this item
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.
- DNase, RNase, and Protease tested
TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.
Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought