Order Entry
ContactUsLinkComponent
TRIS HCl
Tris HCl Buffer Solution, Quality Biological
Catalog #: 10128-396
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
TRIS HCl
Tris HCl Buffer Solution, Quality Biological
Catalog #: 10128-396
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • pH:
    8
  • Description:
    Tris HCl, 1 M
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Test description:
    DNase, RNase and protease tested
  • Shelf life:
    36 months
  • pH:
    7.5 ±0.1
  • Shipping temperature:
    Ambient
  • Supplier No.:
    351-007-101

Specifications

About this item

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These solutions of Tris hydrochloride provide convenient and highly purified solutions.

Tris is compatible with many enzymes in molecular biology, for example DNA modifying enzymes, because its buffering range is between 7.1 - 8.9. The lower pH 7.4 can be associated more with maintaining RNA, however, the pH 8.0 works equally as well with both DNA and RNA nucleic acids

Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959).

Use Tris-HCl 1M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Please call a Avantor sales representative if you plan to order more than 100 bottles
Delivery: Tris-HCl pH 7.0 1M, and Tris-HCl pH 8.8 1.5M require Special Production, 2-4 weeks for delivery
Caution: Research use only.
Customers Who Bought This Also Bought