To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:SOC medium
- Size:10 ml
- Sterility:Autoclave sterilized
- Shipping conditions:Ambient temperature
- Shelf Life:24 months
- Supplier No.:340-031-671
Specifications
About this item
SOC medium is a ready-to-use solution, useful in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli.
- Without magnesium chloride, needs to be added before use.
SOC is a microbial growth medium used for the transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and glucose in a low-salt formulation.
More specifically SOC Medium can be used: in the propagation of DH-alpha (of E. coli strains) primarily in the recovery step of Escherichia coli competent cell transformations, and to maximize the transformation efficiency of competent cells.
Recommendation
Immediately before use, add 2M magnesium chloride to a final concentration of 20mM (10ml MgCl2/L).
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99