Sabouraud Dextrose (SabDex) Agar Blue, Hardy Diagnostics
Catalog #: CA89429-348
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Plate
- Description:SabDex Agar Blue USP
- Size:15x100mm
- Storage temperature:2-8C
- Cat. no.:CA89429-348
- Fill Volume:26 ml
- Application:For the cultivation of fungi.
- Supplier No.:W82
Specifications
About this item
Sabouraud Dextrose (SabDex) Agar, Blue, not for human diagnostic use, 26ml fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
This product is for laboratory use only; not for human diagnostic use. See VWR catalog number 89407-894 for Sabdex Agar that is intended for human diagnostic use.
This product contains an inert blue dye to facilitate differentiation from other media that would otherwise have an similar color (such as Tryptic Soy Agar).
(Sabourauds Dextrose Agar) For the cultivation of fungi.
This product contains an inert blue dye to facilitate differentiation from other media that would otherwise have an similar color (such as Tryptic Soy Agar).
Certifications: USP
Customers Who Bought This Also Bought