Order Entry
ContactUsLinkComponent
Liquid biological indicator
SporAmpule® Liquid Biological Indicator
Catalog #: CA89422-664
CAS Number:  
Liquid biological indicator
SporAmpule® Liquid Biological Indicator
Catalog #: CA89422-664
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    20 per package
  • Description:
    SporAmpule® Liquid biological indicator, for steam sterilization
  • Storage temperature:
    2...8 ℃
  • Supplier No.:
    BST20

Specifications

About this item

SporAmpule®, liquid biological indicator, for steam sterilization, 100 per package, by Hardy Diagnostics.

An error-free, BI (Biological Indicator) system certified for use with steam sterilization processes. After processing in a normal load, the SporAmpule® is incubated at 55 to 60 °C for 48 hours. A purple to yellow color change and visible turbidity of a processed SporAmpule® indicates sterilizer failure.

Industries: Breweries, wineries and distilleries, cannabis QA Testing, clinical, cosmetics, environmental, food safety, pharmaceutical manufacturing.

Customers Who Bought This Also Bought