Tryptic Soy Broth (TSB), Hardy Diagnostics
Catalog #: CA89408-048
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Serum bottle with needle-port septum
- Description:TSB, USP
- Size:100 mL
- Storage conditions:–2-30°C
- Minimum Order Quantity:—
- Cat. no.:CA89408-048
- Supplier No.:U46
Specifications
About this item
For the general cultivation of microorganisms.
Delivery: Ships Ground Only.
Customers Who Bought This Also Bought