Lok-Tight™ Contact Plates, Hardy Diagnostics
Catalog #: CA89408-026
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:Tryptic Soy Agar (TSA) with Lecithin and Tween® 80
- Size:15 x 65 mm
- Storage conditions:2-8°C
- Minimum Order Quantity:—
- Cat. no.:CA89408-026
- Supplier No.:P34
Specifications
About this item
An added measure to ensure sterility in your environmental testing program.
- Friction lid design keeps lids in place
- Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
- Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
Hardy Diagnostics contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.
Customers Who Bought This Also Bought