Order Entry
ContactUsLinkComponent
Emmon's Sabdex Agar
Sabdex (Sabouraud Dextrose) Agar
Catalog #: CA89407-894
CAS Number:  
Emmon's Sabdex Agar
Sabdex (Sabouraud Dextrose) Agar
Catalog #: CA89407-894
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    5.6 ±0.2 at 25 °C
  • Packaging:
    Plate
  • Description:
    Sabouraud Dextrose Agar (SabDex), USP
  • Fill Volume:
    26 ml
  • Storage Temperature:
    2...8 °C
  • Application:
    Breweries, wineries and distilleries, cannabis QA testing, cosmetics, environmental, food safety
  • Dimensions:
    15×100 mm
  • Supplier No.:
    W70

Specifications

About this item

Sabouraud Dextrose Agar, Sabouraud Dextrose Broth, and Sabouraud Dextrose Agar, Emmons are recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.

Ideal for environmental monitoring in a clean room or clean bench setting.

Sabouraud Dextrose Agar with Chloramphenicol, Sabouraud Dextrose Agar with Chloramphenicol and Gentamicin, and Sabouraud Dextrose Agar with Chloramphenicol and Tetracycline are recommended for the selective isolation of fungi and yeasts from clinical and nonclinical specimens.

Caution: Not intended to be used for the diagnosis of human disease.
Customers Who Bought This Also Bought