OxiStrips™, Oxidase Strips, Hardy Diagnostics
Catalog #: CA89407-796
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Paper Strips in Vial
- Size:25 preparations
- Storage temperature:15-30°C
- Cat. no.:CA89407-796
- Supplier No.:Z93
Specifications
About this item
Paper strips for testing the oxidase reaction, contains tetramethyl-p-phenylenediamine for a more sensitive test, simply place a drop of water on the strip, then rub on bacteria, positives turn dark purple within 15 seconds, room temperature storage, one year shelf life from date of manufacture, multiple tests can be performed on one strip
Plastic handle protects hands from active ingredient.
Customers Who Bought This Also Bought