Fluid D, USP
Catalog #: CA89407-538
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Glass bottle with needle port septum
- Size:100 ml
- Storage Temperature:–2...30 °C
- Supplier No.:U110
Specifications
About this item
Fluid D is a rinsing and diluting fluid for use in USP testing protocols.
- Rinsing and diluting fluid
- Contains peptic digest of animal tissue and polysorbate 80
A rinsing and diluting fluid for use in USP testing protocols. pH 7.1. Contains peptic digest of animal tissue and polysorbate 80.
Customers Who Bought This Also Bought