Tryptic Soy Agar (TSA), Soybean Casein Digest Agar, Hardy Diagnostics
Catalog #: CA89407-290
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Plate
- Description:TSA, with Lecithin and Tween® 80
- Size:15 x 100 mm
- Storage temperature:2-8°C
- Volume:20 ml
- Supplier No.:G41
Specifications
About this item
For use as a general growth medium for the isolation and cultivation of microorganisms.
- General growth medium
- For pure cultures
This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms.
Customers Who Bought This Also Bought