Order Entry
ContactUsLinkComponent
Peptone Water
Dilu-Lok™ Dilution Vial with Hinged Cap
Catalog #: CA89407-108
CAS Number:  
Peptone Water
Dilu-Lok™ Dilution Vial with Hinged Cap
Catalog #: CA89407-108
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    50/pack
  • Description:
    Peptone water 0.1%
  • Storage temperature:
    –2-30°C
  • Fill Volume:
    99 ml
  • Final Volume:
    145 ml
  • Supplier No.:
    D299

Specifications

About this item

The Dilu-Lok™ eliminates the time and money spent on preparing your dilution vials! Our pre-filled dilution vial is designed especially for the food, dairy, pharmaceutical, and water industries. The vials are filled to 25, 90, or 99 ml with a variety of buffering and enrichment solutions. Each lot is meticulously tested for sterility and fill volume.

  • Heavy duty: Vials are made of sturdy polypropylene with a leak-resistant seal that holds up even during air transport
  • Leak resistant: Re-engineered for an improved seal, easy-to-remove, perforated, shrink wrap ensures product integrity
  • Optically clear: vials are made of clarified polypropylene for clear viewing

Large opening: A wide 45mm opening accommodates most sample sizes. Each vial is clearly marked with a 90 and 99 ml fill line.
One-handed: Flip-top lid allows for easy one-handed opening and closing to make pipetting easier. Saves unnecessary motion! No awkward screw caps or plastic strips to tear off.
Variety: Available with peptone water, buffered peptone water, Butterfield's buffer, deionized water, phosphate buffer with MgCl, and an empty sterile container.

Industry: Breweries, wineries and distilleries, canabis QA testing, cosmetics, environmental, food safety

Customers Who Bought This Also Bought