Order Entry
ContactUsLinkComponent
Agar
CRITERION™ Agar, Plant Tissue Culture, Grade Course, Hardy Diagnostics
Catalog #: CA89406-838
CAS Number:  
CRITERION™ Agar, Plant Tissue Culture, Grade Course, Hardy Diagnostics
Catalog #: CA89406-838
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    500 g
  • Storage temperature:
    –2…+30 °C
  • Cat. no.:
    CA89406-838
  • Supplier No.:
    C8941

Specifications

About this item

CRITERION™ Agar, Plant Tissue Culture, Grade Purified, used for culturing plant tissues.

  • Wide mouth opening reduces dust and allows easy scooping
  • Convenient handgrip with 'no slip' finger holds
  • Stackable containers
  • High solubility
  • Custom and bulk products available

Plant tissue culturing is the process of propagating plants in culture media, usually to control the environment in such a way to achieve strong seedlings for mass production. Also used when companies genetic engineer plants and want to control their environment. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.

Certifications: BSE Free
Customers Who Bought This Also Bought