Order Entry
ContactUsLinkComponent
Baird Parker Agar Base
Baird Parker Agar Base, CRITERION™, Hardy Diagnostics
Catalog #: CA89405-106
CAS Number:  
Baird Parker Agar Base
Baird Parker Agar Base, CRITERION™, Hardy Diagnostics
Catalog #: CA89405-106
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Wide-Mouth Bottle
  • Size:
    500 g
  • Storage temperature:
    –2-30°C
  • Cat. no.:
    CA89405-106
  • Supplier No.:
    C5081

Specifications

About this item

CRITERION™ Baird-Parker Agar Base is used with Egg Yolk Tellurite Enrichment as a selective medium for the detection and enumeration of coagulase-positive staphylococci from food samples

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Egg Yolk Tellurite Emulsion, product 89407-702, is required for this meduim. Dehydratedculture media with engineered container.

Certifications: BSE Free
Customers Who Bought This Also Bought